Learn More
Amberlite™ FPC3500(H), ion exchange resin, Thermo Scientific™
550.00 NOK - 1470.00 NOK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442547
|
Thermo Scientific Alfa Aesar
045903.30 |
250 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15452547
|
Thermo Scientific Alfa Aesar
045903.A1 |
1 kg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Suitable for the neutralization of strong bases, the recovery of metallic ions, and the isolation and concentration of antibiotics, basic amino acids, enzymes, and peptides