Learn More
Amberlite™ FPC3500 Cation, for Ion Chromatography, Carboxylic Functionality, Hydrogen form (size 0.45-0.65mm), Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
Specifications
Specifications
| Physical Form | Solid |
| CAS | 950182-46-4 |
| Packaging | Plastic powder jar |
| MDL Number | MFCD08460528 |
| Color | White |
| Quantity | 100 g |