missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom™, 50WX2, 200-400 mesh, H-form, Thermo Scientific™

Product Code. 17515625 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 17515625

Brand: Thermo Scientific Acros 473035000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 69011-20-7, 7732-18-5
Molecular Formula C28H30
Molecular Weight (g/mol) 0.00
MDL Number MFCD00132722
InChI Key NWUYHJFMYQTDRP-UHFFFAOYSA-N
IUPAC Name 1,2-diethenylbenzene; 1-ethenyl-2-ethylbenzene; ethenylbenzene
SMILES *

Specifications

Physical Form Beads
CAS Min % 18%, 74%
CAS Max % 26%, 82%
Ion Exchange Capacity ≥0.6 eq/L (H form)
Particle Size 200 to 400 mesh, ≤15 % (< 50 μm), ≤15 % (> 210 μm)
Packaging Plastic Bottle
Water Retention Capacity 74 to 82 % (H form)
Color White to Yellow to Orange
Quantity 500 g
Solubility Information Insoluble in water
Chemical Name or Material AmberChrom™
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.