Learn More
Emma Safety Footwear Amber Safety Shoes
Product Code. 15939101
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
Unit Size:
1 pair
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15939101 37 1 pair
15919101 35 1 pair
15929101 36 1 pair
15949101 38 1 pair
15959101 39 1 pair
15969101 40 1 pair
15979101 41 1 pair
15989101 42 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15939101 missing translation for 'mfr' Emma Safety Footwear missing translation for 'supplierNo' MM94251637
Supremely comfortable, sculpted ladies safety shoe. Features SmartTecAir™ technology to remove moisture and perspiration instantly.
  • S1P-Safety shoe with sole plate for dry work
  • GripForce™ Easy Twist
  • Microfiber construction
  • ESD: Electrostatic Discharge
  • Chrome-free lining
  • Hydro-Tec™ Sanitized Silver
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Amber
Liner Material Hydro-Tec™ Sanitized Silver
Material Microfiber
Size 37
Toe Material Steel