Learn More
Fisherbrand™ Amber HDPE Narrow-Mouth Bottles
Product Code. 12642425 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Neck Finish:
24-400
28-400
38-430
Unit Size:
Pack of 12
Pack of 24
Pack of 48
Pack of 6
Pack of 72
Product Code. Capacity (Metric) Neck Finish unitSize
12642425 1000 mL 38-430 Pack of 6
11937944 125 mL 24-400 Pack of 72
12622425 125 mL 24-400 Pack of 12
11947944 250 mL 24-400 Pack of 72
12612425 250 mL 24-400 Pack of 12
11947924 500 mL 28-400 Pack of 48
12632425 500 mL 28-400 Pack of 12
11927914 1000 mL 38-430 Pack of 24
8 options
Product Code. 12642425
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Amber polypropylene screw caps
Protect light-sensitive materials during sampling, shipping or storage
  • Amber high-density polyethylene
  • Available in various capacities
Fisherbrand HDPE containers can be used for long-term storage, but direct UV exposure should be avoided.
Compliance
Meet U.S. Pharmacopoeia 661 (Containers, Light Transmission).
TRUSTED_SUSTAINABILITY
Specifications
Capacity 32 oz., 1000 mL
Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Amber
Material HDPE
Neck Finish 38-430
Certifications/Compliance U.S. Pharmacopoeia 661
Closure Material Polypropylene
Closure Size 38-430 mm
Closure Type Screw Cap