Learn More
Fisherbrand™ Amber HDPE Narrow-Mouth Bottles
GREENER_CHOICE
Product Code. 12632425 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
1000 mL
Neck Finish:
24-400
28-400
38-430
Unit Size:
Pack of 12
Pack of 24
Pack of 48
Pack of 6
Pack of 72
Product Code. 12632425
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Amber polypropylene screw caps
Protect light-sensitive materials during sampling, shipping or storage
  • Amber high-density polyethylene
  • Available in various capacities
Fisherbrand HDPE containers can be used for long-term storage, but direct UV exposure should be avoided.
Compliance
Meet U.S. Pharmacopoeia 661 (Containers, Light Transmission).
TRUSTED_SUSTAINABILITY
Specifications
Capacity 17 oz., 500 mL
Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Amber
Material HDPE
Neck Finish 28-400
Certifications/Compliance U.S. Pharmacopoeia 661
Closure Material Polypropylene
Closure Size 28-400 mm
Closure Type Screw Cap