Learn More
Lenz Laborglasinstrumente™ Amber Erlenmeyer Flasks with Ground Joint
Click to view available options
:
Capacity: 100mL
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 4797 |
| Capacity (Metric) | 100 mL |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | Erlenmeyer Flask, 100mL, 64 mm dia., Amber, Duran Glass, 29/32 NS Socket, with ground joint, Inscription Label |
| Diameter (Metric) Outer | 64 mm |