Learn More
BRAND™ Amber Conical Ground Joint Stoppers
Product Code. 15690479 Shop All Brand Products
Change view
Click to view available options
Size:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 24/29
Unit Size:
Each
Product Code. Size unitSize
15690479 NS 19/26 Each
15660479 NS 10/19 Each
15670479 NS 12/21 Each
15680479 NS 14/23 Each
15600489 NS 24/29 Each
5 options
Product Code. 15690479
Brand: BRAND™ 144674
Only null left
Add to basket
Add to basket
Boro 3.3 honed stoppers
TRUSTED_SUSTAINABILITY
Specifications
Description Honed conical joint stopper
Quantity 1
Size NS 19/26
Material Borosilicate Glass