Learn More
Heathrow Scientific™ Aluminum Test Tube Basket
Product Code. 12327569 Shop All Thermo Scientific CO₂ Incubators Products
Change view
Click to view available options
Dimensions (L x W x H):
152 x 152 x 152 mm
256 x 152 x 152 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) Unit Size
12327569 256 x 152 x 152 mm Each
12317569 152 x 152 x 152 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12327569 Supplier Heathrow Scientific™ Supplier No. HS20341B
  • ONLINE EXCLUSIVE PRICE
    364.00 NOK
    297.89 NOK / Pack of 1
    Save 66.11 NOK
    18% Off
Estimated Shipment: 29-12-2026
Wash and store test tubes. Heathrow Scientific™ Aluminum Test Tube Basket can withstand repeated exposure to autoclaves.
  • Constructed of lightweight aluminum
  • Ideal for washing and storing test tubes
  • Withstands repeated autoclaving and use in the lab
Specifications
Autoclavable Autoclavable
Color Silver
Holds Test Tubes
Material Aluminum
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
For Use With (Application) Washing, Autoclaving, Storing Test Tubes
Dimensions (L x W x H) 256 x 152 x 152 mm
Product Type Basket for Test Tubes
Height (Metric) 15.2 cm
Array 1 x 1
Width (Metric) 15.2 cm