Learn More
VACUUBRAND™ Aluminum T-piece With Small KF Flanges
Product Code. 10589911
Change view
Click to view available options
Diameter (Metric) Inside:
10/10/10 mm
16/16/16 mm
25/25/25 mm
40/40/40 mm
Size:
30 x 60 mm
40 x 80 mm
50 x 100 mm
65 x 130 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside Size Unit Size
10589911 40/40/40 mm 65 x 130 mm Each
10039560 10/10/10 mm 30 x 60 mm Each
10273371 16/16/16 mm 40 x 80 mm Each
10450273 25/25/25 mm 50 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10589911 Supplier VACUUBRAND™ Supplier No. 20669506
Estimated Shipment: 08-12-2026