missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Sulfate Hydrate ACS MP Biomedicals

Product Code. 11440482 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 11440482

Brand: MP Biomedicals 0219140390

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17927-65-0
MDL Number MFCD00149136
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18
PubChem CID 22377415
ChEBI CHEBI:74779

Specifications

Quantity 500 g
Assay Percent Range 98 to 102%
Molecular Formula Al2O12S3
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
SMILES [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
IUPAC Name dialuminium(3+) trisulfate
Molecular Weight (g/mol) 342.13
Formula Weight 342.15 (Anhydrous)
Grade ACS Reagent
Chemical Name or Material Aluminum Sulfate Hydrate, Hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.