Learn More
Brady™ Aluminum Sign: Water fog applicator - ISO 7010
Product Code. 15644197
Click to view available options
Width (Metric):
100 mm
150 mm
200 mm
Height (Metric):
100 mm
150 mm
200 mm
Unit Size:
Each
Product Code. 15644197
Brand: Brady™ 135954
Only null left
Add to basket
Add to basket
Sign compliant to ISO 7010.
  • Aluminum
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number F/F011/NT-ALU-200X200/1-B
Material Aluminum
Quantity 1 Ea.
Height (Metric) 200 mm
Thickness 1.016 mm
Width (Metric) 200 mm
Temperature Range (Metric) -40°C to 140°C
Shape Square
Languages Global
Color White on Red