Learn More
Macherey-Nagel™ ALUGRAM™ Alox N/UV254 Aluminum Sheets
Product Code. 11386474 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
5 x 20 cm
20 x 20 cm
Quantity:
25 Pk.
50 Pk.
Unit Size:
Pack of 25
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Quantity Unit Size
11386474 20 x 20 cm 25 Pk. Pack of 25
10659581 5 x 20 cm 50 Pk. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11386474 Supplier Macherey-Nagel™ Supplier No. 818023
  • ONLINE EXCLUSIVE PRICE
    1995.00 NOK
    1381.51 NOK / Pack of 25
    Save 613.49 NOK
    30% Off
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Designed for use in thin layer chromatography. Macherey-Nagel™ ALUGRAM™ Alox N/UV254 Aluminum Sheets include aluminum oxide layers and a fluorescent indicator at short-wave UV (254nm). Activating the layers before use is recommended.
ALUGRAM™ Alox N/UV254 are aluminum sheets with aluminum oxide layers for use in TLC. They feature an inert organic binder and a manganese-activated zinc silicate indicator.
  • Recommended application: terpenes, alkaloids, steroids, aliphatic and aromatic compounds
  • Activate the aluminum oxide layers before use by heating 10 minutes at 120™C
Specifications
Material Aluminum Sheets Alugram Alox N
Pore Size 60 Å
Quantity 25 Pk.
Type TLC Plate
Fluorescent Indicator UV254
For Use With (Application) Terpenes, Alkaloids, Steroids, Aliphatic and Aromatic Compounds
Specific Surface ∼200 m2/g
Thickness (Metric) 0.2 mm
Dimensions (L x W) 20 x 20 cm