Learn More
Brady™ Aluminum with Polyester Overlaminate Transport Signs
Product Code. 11770244
Change view
Click to view available options
Displayed Text:
1.5 1
1.6 1
3
4
Unit Size:
Each
Product Code. Displayed Text unitSize
11770244 3 Each
11780234 1.5 1 Each
11700254 4 Each
11710244 1.6 1 Each
4 options
Product Code. 11770244
Brand: Brady™ 223603
Only null left
Specifications
Alternate Part Number ADR 3B-297*297-B7525
Material Aluminum
Quantity 1 Ea.
Height (Metric) 297 mm
Thickness 1.016 mm
Width (Metric) 297.00 mm
Temperature Range (Metric) -40 °C-140 °C
Shape Diamond
Displayed Text 3
Color White on Red