Learn More
Brady™ Aluminum with Polyester Overlaminate Transport Signs
Specifications
Specifications
| Alternate Part Number | ADR 1.6-297*297-B7525 |
| Material | Aluminum |
| Quantity | 1 Ea. |
| Height (Metric) | 297 mm |
| Thickness | 1.016 mm |
| Width (Metric) | 297.00 mm |
| Temperature Range (Metric) | -40 °C-140 °C |
| Shape | Diamond |
| Displayed Text | 1.6 1 |
| Color | Black on Orange |