Learn More
Brady™ Aluminum with Polyester Overlaminate Radioactive Warning Sign
Click to view available options
Height (Metric):
315 mm
Unit Size:
Each
Specifications
Specifications
| Material | Aluminum, Polyester |
| Quantity | 1 Ea. |
| Description | Aluminum with Polyester Overlaminate |
| For Use With (Application) | Used for indoor and outdoor safety signs, signal radioactive materials |
| Height (Metric) | 315 mm |
| Width (Metric) | 315 mm |
| Shape | Triangular |
| Displayed Text | Warning of Radioactive materials or radiation ioniserenden |
| Product Type | Radioactive Warning Sign |