Learn More
Nickel Electro™ Aluminum Plate Sandbath Kit for use with HP1-1 Hotplates
Product Code. 11419499 Shop All Nickel-Electro Products
Change view
Click to view available options
Height (Metric):
27 mm
Unit Size:
Each
Product Code. Height (Metric) unitSize
11419499 27 mm Each
1 options
Product Code. 11419499
Brand: Nickel Electro™ SB11
Only null left
Add to basket
Add to basket
Nickel Electro™ Aluminum Plate Sandbath Kit converts flat top HP1-1 hotplates into sandbath for immersion of small samples.
  • Aluminum
  • Used to covert hotplates into sand baths
Specifications
Height (Metric) 27 mm