Learn More
Cole Parmer™ Aluminum Pan Liners
Product Code. 10472772 Shop All Cole-Parmer Products
Click to view available options
Quantity:
500/Cs.
Unit Size:
Pack of 500
Product Code. 10472772
Brand: Cole Parmer™ 0101824
This item requires a price update prior to being ordered.
Request a Price for this item here.
Designed for moisture determination balances.
  • Suitable for use with moisture determination balances.
  • Disposable.
  • Oil-free.
  • Ideal for general purpose weighting and dispensing.
  • Compatible with both open and enclosed balances.
  • Slightly tapered for stacking.
Specifications
Disposable Single-use
Material Aluminum
Shape Round
Type Weighing Boat
Quantity 500/Cs.
Width (Metric) 4 in.
Height (English) 0.313 in.
Depth (Metric) 0.8 cm
Diameter (English) 4 in.
Diameter (Metric) 10.2 cm
Weight (Metric) 2.5 G