Learn More
Aluminum Oxide Substrate, 12.7 x 12.7 x 1mm, Polished One Side, M Plane, Thermo Scientific™
Product Code. 11369338
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
Unit Size:
Each
Pack of 5
Product Code. Quantity unitSize
11369338 1 Ea. Each
11379338 5 Ea. Pack of 5
2 options
Product Code. 11369338
Brand: Thermo Scientific Alfa Aesar 043246.KT
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Aluminum Oxide Substrate, M Plane
Name Note Polished One Side
Quantity 1 Ea.
Molecular Formula Al2O3
MDL Number MFCD00003424
Solubility Information Soluble in hexane and toluene. Insoluble in water and diethyl ether.