Learn More
Haldenwanger™ Aluminum Oxide Combustion Boats
Description
Aluminum oxide boats of great hardness, strength and chemical resistance. Suitable for continuous use up to 1, 700°C to 1, 750°C.
Specifications
Specifications
| Type | Combustion Boat |
| Dimensions (L x W x H) | 200 x 15 x 15 mm |
| For Use With (Application) | For continuous use up to 1700 to 1750°C. |
| Height (Metric) | 15 mm |
| Material | Aluminum Oxide |
| Quantity | 1 Ea. |