Description
- Smooth-sided, flat bottom, multipurpose dish
- Oil-free and non-coated
- Aluminum Oil Free
- Disposable
Specifications
Specifications
| Disposable | Single-use |
| Material | Aluminum |
| Shape | Round |
| Type | Weighing Dish |
| For Use With (Application) | Universal balances |
| Height (Metric) | 6 mm |
| Depth (Metric) | 76.2 mm |
| Diameter (Metric) | 64 mm |
| Weight (Metric) | 1.3 g |