Learn More
Sampling Systems™ Aluminum Multilevel Slot Sampler
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
880 mL
1.7 L
2.9 L
Length (Metric):
900 mm
1500 mm
2500 mm
Includes:
3 Slots
5 Slots
7 Slots
Unit Size:
Each
Description
- Made from aluminum
- Cross section (Composite sample)
- Suitable for taking large volume samples
- aluminum samplers are not suitable for use with GMP products
Specifications
Specifications
| Material | Aluminum |
| Description | Slot Sampler |
| Includes | 3 Slots |
| Capacity (Metric) | 880 mL |
| Diameter (Metric) Outer | 50 mm |
| Length (Metric) | 900 mm |