Learn More
Aluminum magnesium isopropoxide, 99.95+% (metals basis)
Product Code. p-7050712
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
Product Code. 11313738
Brand: Thermo Scientific Alfa Aesar 039320.04
Request Bulk or Custom Format
CAS: 69207-83-6
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69207-83-6
MDL Number MFCD00070426
Specifications
Melting Point 40°C
Boiling Point 114°C (0.2 mmHg)
Quantity 2 g
UN Number UN3181
Sensitivity Moisture sensitive
Formula Weight 550.53
Percent Purity ≥99.95%
Physical Form Crystalline Low Melting Solid
Assay (metals basis)
Chemical Name or Material Aluminum magnesium isopropoxide
compliance-icons compliance-icons
Product Identifier
  • Aluminum magnesium isopropoxide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Flammable solid Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .