Learn More
Bochem™ Aluminum Laboratory Pot With Lid
Brand: Bochem™ 8568
Additional Details : Weight : 0.80000kg
Specifications
5.2L | |
240mm | |
Laboratory Pot | |
With lid |
Yes | |
Aluminum | |
140mm |