Learn More
Welch™ Aluminum KF Inner Centering with Viton O-ring
Product Code. 10152522 Shop All Welch Ilmvac Products
Change view
Click to view available options
For Use With:
Separator 16 DN
Separator 25 DN
Separator 40 DN
Unit Size:
Each
Product Code. For Use With unitSize
10152522 Separator 25 DN Each
10680343 Separator 16 DN Each
10090992 Separator 40 DN Each
3 options
Product Code. 10152522
Brand: Welch™ 701093
Only null left
Add to basket
Add to basket
Designed for use with CRVpro Rotary Vane Vacuum Pumps. Welch Ilmvac™ Aluminum KF Inner Centering Rings feature a replaceable Fluoroelastomer O-ring seal.
  • Stainless steel centering ring with Viton O-ring
  • Replaceable Fluoroelastomer O-ring seal
  • Centering ring assemblies are placed between two ISO flanges with matching outer diameters
  • he widest portion of the centering ring rests inside a centering ring locater groove (blank flange) or counterbore (bored flange) on the Kwik-Flange™
  • The O-ring rests on the flat polished surface outside the locater groove or counterbore
TRUSTED_SUSTAINABILITY
Specifications
For Use With Separator 25 DN
Material Aluminum
Quantity 1 Each
Type KF Inner Centering