Learn More
Welch™ Aluminum KF Inner Centering with Buna N O-ring
Product Code. 10640153 Shop All Welch Ilmvac Products
Click to view available options
Diameter (Metric) Outside:
16 mm
25 mm
40 mm
Unit Size:
Each
Product Code. 10640153
Brand: Welch™ 701083
Only null left
Specifications
Diameter (Metric) Outside 25 mm
For Use With Separator 25 DN
Material Aluminum
Quantity 1 Each
Type KF Inner Centering