Learn More
Brady™ Aluminum: ISO Safety Sign - Fire alarm call point
Product Code. 15644417 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
100 mm
200 mm
Height (Metric):
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Height (Metric) Unit Size
15644417 200 mm 200 mm Each
15634417 100 mm 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15644417 Supplier Brady™ Supplier No. 817689
  • ONLINE EXCLUSIVE PRICE
    135.00 NOK
    129.54 NOK / Pack of 1
    Save 5.46 NOK
    4% Off
In Stock
Sign compliant to ISO 7010.
  • Aluminum
  • For indoor and outdoor use
Specifications
Certifications/Compliance ISO 7010
Alternate Part Number PIC F005-200x200-AL-CRD/1
Material Aluminum
Quantity 1 Ea.
For Use With (Application) Indoor and Outdoor
Height (Metric) 200 mm
Thickness 1.016 mm
Width (Metric) 200 mm
Temperature Range (Metric) -40°C to 140°C
Shape Square
Languages Global
Color White on Red