Learn More
Cole Parmer™ Aluminum General Purpose Weighing Dishes
Product Code. p-7161089 Shop All Cole-Parmer Products
Change view
Click to view available options
Capacity (Metric):
20 mL
75 mL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15487903 20 mL Pack of 100
15456083 75 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15487903 Supplier Cole Parmer™ Supplier No. 0101860
Ideal for sampling, weighing, dispensing, storage, environmental clean-up, food processing, evaporating, or general laboratory use.
  • Disposable.
  • Suitable for a variety of laboratory weighing, dispensing, processing uses.
  • All pans are contaminate and oil-free.
  • Dishes are stackable and made from light-gauge aluminum.
Specifications
Bottom Shape Flat
Capacity (Metric) 20 mL
Disposable Single-use
Material Aluminum
Shape Round
Type Weighing Dish
Quantity 100/Pk.
Height (English) 0.5 in.
Diameter (English) 2 in.