Learn More
Brady™ Aluminum Evacuation Panel Safety Signs
Product Code. 11720973
Change view
Click to view available options
Width (Metric):
148 mm
200 mm
400 mm
Style:
with Down Arrow
with Left Arrow
with Right Arrow
with Up Arrow
Displayed Text:
Exit and emergency exit
Unit Size:
Each
Product Code. Width (Metric) Style Displayed Text unitSize
11720973 148 mm with Left Arrow Exit and emergency exit Each
11750833 148 mm with Down Arrow Exit and emergency exit Each
11780833 200 mm with Down Arrow Exit and emergency exit Each
11700983 148 mm with Right Arrow Exit and emergency exit Each
11730843 148 mm with Up Arrow Exit and emergency exit Each
11760843 400 mm with Up Arrow Exit and emergency exit Each
6 options
Product Code. 11720973
Brand: Brady™ 811161
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Material Aluminum
Quantity 1 Ea.
Style with Left Arrow
For Use With (Application) for Pipe Markers and Safety Signs
Height (Metric) 297 mm
Max. Temperature (Metric) 120°C
Width (Metric) 148 mm
Displayed Text Exit and emergency exit
Product Type Evacuation Panel Safety Sign
Color White on Green