Learn More
Bochem™ Aluminum Epoxy Lab jack
Description
- Improved scissor technology with reduced tolerances
- Ergonomic knob, rounded and user friendly
- Scissor-construction made of 18/10-stainless steel
- Manufactured according to DIN 12897
- Maximal dynamic operating load (Kg max. dyn.) and maximal static operating load (Kg max. stat.) norms according to DIN 12897
- Smaller models (up to 240x240mm; Typ 1) have a simple hand-operated adjusting wheel
- Bigger models (from 300x300mm; Typ 2) use of included ratchet, which connects with a special adjusting wheel, is recommended
Specifications
Specifications
| Material | Aluminum Epoxy |
| Height (Metric) | 60 to 275 mm |
| Length (Metric) Base | 240 mm |
| Product Type | Lab Jack |
| Quantity | 1 Ea. |