Learn More
Nickel Electro™ Aluminum Dissection Dishes
Product Code. 12780026 Shop All Nickel-Electro Products
Change view
Click to view available options
Height (Metric):
50 mm
55 mm
Length (Metric):
260 mm
310 mm
450 mm
Width (Metric):
30 mm
200 mm
360 mm
Weight (Metric):
0.2 kg
0.7 kg
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Length (Metric) Width (Metric) Weight (Metric) unitSize
12780026 55 mm 260 mm 360 mm 0.2 kg Each
12770026 50 mm 310 mm 200 mm 0.2 kg Each
12790026 50 mm 450 mm 30 mm 0.7 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12780026 Supplier Nickel Electro™ Supplier No. 626
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Hold small specimens while transporting. Nickel Electro™ Aluminum Dissecting Dish is useful for working with and transporting small specimens.
  • Aluminum
  • Shallow form
  • One piece pressing
  • Useful when working with and transporting small specimens
Specifications
Product Type Dissection Dish
Autoclavable Autoclavable
Material Aluminum
Product Line Dissection Dish
Shape Rectangular
Length (Metric) 260 mm
Width (Metric) 360 mm
Height (Metric) 55 mm
Weight (Metric) 0.2 kg