Learn More
Thermo Scientific Chemicals Aluminum Dish, Flat Bottom & Straight Sides;Dia (mm), 76;Wall Ht (mm), 25
Product Code. 11353458
Change view
Click to view available options
Diameter (Metric):
51 mm
76 mm
90 mm
126 mm
204 mm
Height (Metric):
12.5 mm
25 mm
51 mm
No. per Pack:
2/Pk.
6/Pk.
Unit Size:
Pack of 2
Pack of 6
Product Code. Diameter (Metric) Height (Metric) No. per Pack unitSize
11353458 76 mm 25 mm 6/Pk. Pack of 6
11490191 51 mm 12.5 mm 2/Pk. Pack of 2
11329107 51 mm 12.5 mm 6/Pk. Pack of 6
11420201 76 mm 25 mm 2/Pk. Pack of 2
11430201 90 mm 51 mm 2/Pk. Pack of 2
11363458 90 mm 51 mm 6/Pk. Pack of 6
11440201 126 mm 25 mm 2/Pk. Pack of 2
11373458 126 mm 25 mm 6/Pk. Pack of 6
11460201 204 mm 51 mm 2/Pk. Pack of 2
11393458 204 mm 51 mm 6/Pk. Pack of 6
10 options
Product Code. 11353458
Brand: Thermo Scientific Chemicals 039072.UD
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Bottom Shape Flat
Material Aluminum
Shape Flat Bottom and Straight Sides
Type Dish
No. per Pack 6/Pk.
Height (Metric) 25 mm
Diameter (Metric) 76 mm