Learn More
KGW Isotherm™ Dish-Shaped Dewar Flasks
Product Code. 10790972
Change view
Click to view available options
Capacity (Metric):
260 mL
400 mL
800 mL
5300 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10790972 800 mL Each
10139700 400 mL Each
10458551 5300 mL Each
10221461 260 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10790972 Supplier KGW Isotherm™ Supplier No. 1195
  • ONLINE EXCLUSIVE PRICE
    4247.10 NOK
    3730.52 NOK / Pack of 1
    Save 516.58 NOK
    12% Off
View all promotions
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
For the cooling of small samples. KGW Isotherm™ Dish-Shaped Dewar Flasks are suitable as a hot or cold bath.
  • Easy handling
  • Suitable on magnetic stirrer
  • Full silvered
  • Made of borosilicate glass 3.3
  • Polyethylene lid (not included)
Specifications
Cap Lid Type Lid not included
Color Silver
Depth (Metric) Inside 80mm
Diameter (Metric) Outer 170mm
For Use With (Application) Laboratory technique, Medicinal technique
Height (Metric) 125mm
Base Material Borosilicate Glass
Capacity (Metric) 800 mL
Material Aluminum