Learn More
Bochem™ Aluminum Chemical Scoops
Product Code. p-8005443
Change view
Click to view available options
Length (Metric):
135 mm
185 mm
210 mm
430 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10524651 210 mm Each
10718291 430 mm Each
10769461 185 mm Each
10050891 135 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10524651 Supplier Bochem™ Supplier No. 8819
Only null left
Add to basket
Add to basket
Manufactured from aluminum. Bochem™ Aluminum Chemical Scoops comes in many sizes.
  • Chemical scoop
  • Manufactured from aluminum
  • Comes in many sizes
Specifications
Quantity 1 Ea.
Capacity (Metric) 160 mL
Product Type Chemical Scoop
Diameter (Metric) Outer 37mm
Material Aluminum
Length (Metric) 210 mm