Learn More
Bochem™ Aluminum Chemical Scoops
Product Code. 10050891
Click to view available options
Length (Metric):
135 mm
185 mm
210 mm
430 mm
Unit Size:
Each
Product Code. 10050891
Brand: Bochem™ 8816
Only null left
Add to basket
Add to basket
Manufactured from aluminum. Bochem™ Aluminum Chemical Scoops comes in many sizes.
  • Chemical scoop
  • Manufactured from aluminum
  • Comes in many sizes
Specifications
Quantity 1 Ea.
Capacity (Metric) 50 mL
Product Type Chemical Scoop
Diameter (Metric) Outer 24mm
Material Aluminum
Length (Metric) 135 mm