Learn More
Bochem™ Aluminum Chemical Scoop
Product Code. 10796971
Change view
Click to view available options
Capacity (Metric):
10 mL
65 mL
90 mL
240 mL
400 mL
480 mL
900 mL
1.6 L
Length (Metric):
52 mm
145 mm
155 mm
250 mm
270 mm
285 mm
290 mm
310 mm
350 mm
Unit Size:
Each
Product Code. Capacity (Metric) Length (Metric) unitSize
10796971 65 mL 155 mm Each
10655991 400 mL 290 mm Each
10360341 10 mL 52 mm Each
10122221 1.6 L 270 mm Each
10554841 90 mL 250 mm Each
10652882 480 mL 310 mm Each
10662882 900 mL 350 mm Each
10603272 240 mL 145 mm Each
10386001 90 mL 285 mm Each
9 options
Product Code. 10796971
Brand: Bochem™ 8817
Only null left
Add to basket
Add to basket
Manufactured from aluminum, the Bochem™ Aluminum Chemical Scoop – is a perfect addition to your laboratory tools.
  • Lightweight
  • Aluminum
  • Length is total for combined scoop and handle
Specifications
Capacity (Metric) 65 mL
Product Type Chemical Scoop
Material Aluminum
Length (Metric) 155 mm