Learn More
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible
Product Code. p-8008140
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
40 mL
60 mL
80 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10757591 10 mL Each
10383621 25 mL Each
10493711 40 mL Each
10291971 60 mL Each
10445831 80 mL Each
5 options
Product Code. 10757591
Brand: Haldenwanger™ L ALSINTTIEGEL*B/00
Only null left
Add to basket
Add to basket
Use these crucibles for general laboratory applications. Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucibles are resistant to a variety of acids and alkalis.
  • Conical squat form crucibles
  • Aluminum ceramic composition
  • Available in a variety of sizes
  • Aluminum ceramic composition
Specifications
Material Aluminum Ceramic
Capacity (Metric) 10 mL
Product Type Crucible
Shape Conical