Learn More
Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucible
Product Code. 10757591
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
40 mL
60 mL
80 mL
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10757591 10 mL Each
10383621 25 mL Each
10493711 40 mL Each
10291971 60 mL Each
10445831 80 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10757591 Supplier Haldenwanger™ Supplier No. L ALSINTTIEGEL*B/00
  • ONLINE EXCLUSIVE PRICE
    315.00 NOK
    279.08 NOK / Pack of 1
    Save 35.92 NOK
    11% Off
Estimated Shipment: 20-10-2026
Use these crucibles for general laboratory applications. Haldenwanger™ Aluminum Ceramic Conical Squat Form Crucibles are resistant to a variety of acids and alkalis.
  • Conical squat form crucibles
  • Aluminum ceramic composition
  • Available in a variety of sizes
  • Aluminum ceramic composition
Specifications
Material Aluminum Ceramic
Capacity (Metric) 10 mL
Product Type Crucible
Shape Conical