Learn More
Masterflex™ Aluminum 16-Turn Valve Cartridge
Product Code. p-8003441
Click to view available options
Airflow Rate:
0 to 200 mL/min.
1001 to 2500 mL/min.
6201 mL/min.
Waterflow Rate:
0 to 6 mL/min.
30 to 70 mL/min.
650 mL/min.
Unit Size:
Each
Product Code. 15207228
Brand: Masterflex™ 0321862
Only null left
Specifications
Description Range 4
For Use With (Equipment) Aluminum Frame Systems
Type 16-Turn Valve Cartridge
Airflow Rate 1001 to 2500 mL/min.
Waterflow Rate 30 to 70 mL/min.