Learn More
Scharlab ALUMINIUMKLORID-6-HYDRAT PSS 500G
Brand:  Scharlab AL07700500
ALUMINIUMKLORID-6-HYDRAT PSS 500G
Product Code. 11329813
  • 661.50 NOK / 500g
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Aluminium chloride hexahydrate, extra pure, Pharmpur, Ph Eur, BP, USP
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.