Learn More
Bochem™ Aluminum Bosshead
Description
- Aluminum
- Socket screw M8
- 90° angle
Specifications
Specifications
| Type | Bosshead Clamp |
| Material | Aluminum |
| Includes | M8 Socket Screw |
| For Use With (Equipment) | Rods and Stand Bases |
| For Use With (Application) | Holds and Supports Laboratory Apparatus |
| Diameter (Metric) | 16.5 mm |
| Size (Metric) Grip | 16.5 mm |