missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina R, MP EcoChrom™, MP Biomedicals™
Click to view available options
Quantity:
500 g
50 kg
Description
- MP EcoChrom™ Alumina R catalyses a variety of reactions that are useful industrially.
- It is widely used to remove water from gas streams.
- Also used for its hardness and strength.
- It is widely used as an abrasive, including as a much less expensive substitute for industrial diamond.
- Many types of sandpaper use aluminium oxide crystals.
Specifications
Melting Point | ca. 2072 °C |
Color | Pink to White |
pH | 4.3 ± 0.1 |
Physical Form | Free-flowing Powder |
Quantity | 500 g |
Solubility Information | Insoluble in water. |
Formula Weight | 101.96 |
Vapor Pressure | At 20 °C (not applicable) |
Density | 3.94 g/cm3 (at 20°C) |
Chemical Name or Material | Alumina R |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'