Learn More
Macherey-Nagel™ Alugram Aluminum Sheet SILGUR
Product Code. 12794738 Shop All Macherey-Nagel Products
Change view
Click to view available options
Dimensions (L x W):
10 x 20 cm
20 x 20 cm
Unit Size:
Pack of 20
Pack of 25
Product Code. Dimensions (L x W) unitSize
12794738 10 x 20 cm Pack of 20
12750833 10 x 20 cm Pack of 20
12760833 20 x 20 cm Pack of 25
3 options
Product Code. 12794738
Brand: Macherey-Nagel™ 818412
Only null left
Specifications
Particle Size 5 to 17 μm
Pore Size 60 Å
Quantity 20 Pk.
Type Aluminum Sheet
Pore Volume 0.75 mL/g
Specific Surface ∼500 m2/g
Thickness (Metric) 0.20 mm
Dimensions (L x W) 10 x 20 cm