Description
HPTLC aluminum sheets with silanized silica-dimethyl (RP2, C2) modification.
Specifications
Specifications
| Particle Size | 5 to 17 μm |
| Pore Size | 60 Å |
| Quantity | 25 Pk. |
| Type | Aluminum Sheet |
| Fluorescent Indicator | UV254 |
| For Use With (Application) | Active Plant Constituents, Steroids |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Dimensions (L x W) | 20 x 20 cm |