Description
HPTLC aluminum sheets with unmodified silica gel (SiOH) and concentrating zone (kieselguhr, diatomaceous earth or diatomite).
Specifications
Specifications
| Particle Size | 2 to 10 μm |
| Pore Size | 60 Å |
| Quantity | 25 Pk. |
| Type | Aluminum Sheet |
| Fluorescent Indicator | UV254 |
| Pore Volume | 0.75 mL/g |
| Specific Surface | ∼500 m2/g |
| Thickness (Metric) | 0.02 mm |
| Dimensions (L x W) | 10 x 10 cm |