Learn More
Lenz Laborglasintrumente Alu Rings with Hooks
Description
- For cones and sockets to be used with springs
- For the safe connection of conical ground joints
- No breakage of glass hooks
Specifications
Specifications
| Quantity | 10/Pk. |
| For Use With (Application) | Cones and sockets to be used with springs |
| Size | NS 45/40 |