Learn More
Alternate Metals 1 For ICP, SPEX CertiPrep™
Product Code. p-217728 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
500mL
Product Code. 15688335
Brand: SPEX Certiprep WP11
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
AnalyteConcentration
Al20μg/mL
Fe20μg/mL
V20μg/mL
Co10μg/mL
Cu10μg/mL
Mn10μg/mL
Ni10μg/mL
Zn10μg/mL
Be5μg/mL
Sb5μg/mL
Tl5μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
CAS 7697-37-2
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O
Specifications
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Industry Type Wastewater Testing
Quantity 125 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 2% HNO3:
Al    •20μg/mL
Sb    •5μg/mL
Be    •5μg/mL
Co    •10μg/mL
Cu    •10μg/mL
Fe    •20μg/mL
Mn    •10μg/mL
Ni    •10μg/mL
Tl    •5μg/mL
V    •20μg/mL
Zn    •10μg/mL
Chemical Name or Material Alternate Metals 1, For ICP/AA Alternate Water Pollution and Wastewater Standard
Product intended for laboratory use only