Learn More
MP Biomedicals™ Alsever's Solution, 500mL
Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Chemical Name or Material | Alsever's solution |
| Color | Clear, Colorless |
| Concentration | 1X |
| For Use With (Application) | Used as Anti-coagulant/Blood preservative |
| pH | 5.9 to 6.3 |
| Physical Form | Solution |
| Quantity | 500 mL |
| Formulation | Sodium Chloride [NaCl] 4200mg/L, Citric Acid 550mg/L, Dextrose 20500mg/L, Sodium Citrate [2H2O] 8000mg/L |
| Sterility | Sterile |