Learn More
Abnova™ ALS2CR2 Recombinant Protein
Brand:  Abnova™ H00055437-P01.25ug
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16141483
  • 6245.00 NOK / 25µg
Estimated Shipment: 03-07-2024
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
AAH08302
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
71.72
8
Glutathione Sepharose 4 Fast Flow
25 ug
MSLLDCFCTSRTQVESLRPEKQSETSIHQYLVDEPTLSWSRPSTRASEVLCSTNVSHYELQVEIGRGFDNLTSVHLARHTPTGTLVTIKITNLENCNEERLKALQKAVILSHFFRHPNITTYWTVFTVGSWLWVISPFMAYGSASQLLRTYFPEGMSETLIRNILFGAVRGLNYLHQNGCIHRSIKASHILISGDGLVTLSGLSHLHSLVKHGQRHRAVYDFPQFSTSVQPWLSPELLRQDLHGYNVKSDIYSVGITACELASGQVPFQDMHRTQMLLQKLKGPPYSPLDISIFPQSESRMKNSQSGVDSGIGESVLVSSGTHTVNSDRLHTPSSKTFSPAFFSLVQLCLQQDPEKRPSASSLLSHVFFKQMKEESQDSILSLLPPAYNKPSISLPPVLPWTEPECDFPDEKDSYWEF
ALS2CR2/CALS-21/ILPIP/ILPIPA/MGC102916/PAPK/PRO1038
STRADB
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Kinase Assay, Western Blot
55437
STRADB (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
STRADB
Human
Recombinant
Solution