Learn More
Abnova™ ALS2CR2 Recombinant Protein
Brand: Abnova™ H00055437-P01.10ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH08302 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
71.72 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MSLLDCFCTSRTQVESLRPEKQSETSIHQYLVDEPTLSWSRPSTRASEVLCSTNVSHYELQVEIGRGFDNLTSVHLARHTPTGTLVTIKITNLENCNEERLKALQKAVILSHFFRHPNITTYWTVFTVGSWLWVISPFMAYGSASQLLRTYFPEGMSETLIRNILFGAVRGLNYLHQNGCIHRSIKASHILISGDGLVTLSGLSHLHSLVKHGQRHRAVYDFPQFSTSVQPWLSPELLRQDLHGYNVKSDIYSVGITACELASGQVPFQDMHRTQMLLQKLKGPPYSPLDISIFPQSESRMKNSQSGVDSGIGESVLVSSGTHTVNSDRLHTPSSKTFSPAFFSLVQLCLQQDPEKRPSASSLLSHVFFKQMKEESQDSILSLLPPAYNKPSISLPPVLPWTEPECDFPDEKDSYWEF | |
ALS2CR2/CALS-21/ILPIP/ILPIPA/MGC102916/PAPK/PRO1038 | |
STRADB | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Kinase Assay, Western Blot | |
55437 | |
STRADB (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
STRADB | |
Human | |
Recombinant | |
Solution |